Ethyl-3-fluoropyridine-2-carboxylate 95%

Supplier: Apollo Scientific
Warning

PC200030-1G PC200030-500MG PC200030-5G
APOSPC200030-1GEA 65.4 EUR
APOSPC200030-1G APOSPC200030-500MG APOSPC200030-5G
Ethyl-3-fluoropyridine-2-carboxylate 95%
Ethyl-3-fluoropyridine-2-carboxylate
Formula: C₈H₈FNO₂
Melting Pt: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD11616884
CAS Number: 1187732-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR