5-Fluoro-1H-pyrrolo[2,3-b]pyridine
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
5-Fluoro-7-azaindole
PC200049-5G
APOSPC200049-5GEA
145
EUR
APOSPC200049-5G
5-Fluoro-1H-pyrrolo[2,3-b]pyridine
5-Fluoro-1H-pyrrolo[2,3-b]pyridine
Formula:
C7H5FN2 MW: 136,1300 g/mol Melting Pt: 110…113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659678 CAS Number: 866319-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...