Ethyl-4-(trifluoromethyl)-2-pyridinecarboxylate

Supplier: Apollo Scientific
Warning

PC200068-1G PC200068-500MG PC200068-5G
APOSPC200068-1GEA 55.1 EUR
APOSPC200068-1G APOSPC200068-500MG APOSPC200068-5G
Ethyl-4-(trifluoromethyl)-2-pyridinecarboxylate
Ethyl-4-(trifluoromethyl)-2-pyridinecarboxylate
Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12498713
CAS Number: 1171919-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR