4-(Trifluoromethoxy)phenylboronic acid pinacol ester 98%
Supplier: Apollo Scientific
PC200177-25G
APOSPC200177-25GEA
98.6
EUR
APOSPC200177-25G
4-(Trifluoromethoxy)phenylboronic acid pinacol ester 98%
4-(Trifluoromethoxy)phenylboronic acid pinacol ester
Formula:
C₁₃H₁₆BF₃O₃ MW: 288,0706 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05863920 CAS Number: 474709-28-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...