Methyl-5-chloro-6-fluoronicotinate

Supplier: Apollo Scientific

PC200187-1G PC200187-250MG PC200187-5G
APOSPC200187-1GEA 655 EUR
APOSPC200187-1G APOSPC200187-250MG APOSPC200187-5G
Methyl-5-chloro-6-fluoronicotinate
Methyl-5-chloro-6-fluoronicotinate
Formula: C₇H₅ClFNO₂
MW: 189,57 g/mol
Melting Pt: 71…72 °C
Storage Temperature: Ambient
MDL Number: MFCD12498696
CAS Number: 1171918-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR