Methyl-6-(trifluoromethyl)-1H-indol-5-yl ether

Supplier: Apollo Scientific
Danger

Synonyms: 5-Methoxy-6-(trifluoromethyl)-1H-indole

PC200200-1G PC200200-500MG
APOSPC200200-1GEA 386 EUR
APOSPC200200-1G APOSPC200200-500MG
Methyl-6-(trifluoromethyl)-1H-indol-5-yl ether
Methyl-6-(trifluoromethyl)-1H-indol-5-yl ether
Formula: C₁₀H₈F₃NO
MW: 215.18 g/mol
Melting Pt: 104…105 °C
Storage Temperature: Ambient
MDL Number: MFCD11848569
CAS Number: 178896-78-1
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR