1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]azepane
Supplier: Apollo Scientific
PC200216-1G
PC200216-500MG
PC200216-5G
APOSPC200216-1GEA
132
EUR
APOSPC200216-1G
APOSPC200216-500MG
APOSPC200216-5G
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]azepane
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]azepane
Formula:
C₁₂H₁₄ClF₃N₂ MW: 278,71 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13561841 CAS Number: 1219967-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...