Methyl-5-bromo-2-fluoronicotinate

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 5-bromo-2-fluoronicotinate

PC200226-1G PC200226-5G
APOSPC200226-1GEA 74.2 EUR
APOSPC200226-1G APOSPC200226-5G
Methyl-5-bromo-2-fluoronicotinate
Methyl-5-bromo-2-fluoronicotinate
Formula: C₇H₅BrFNO₂
MW: 234,0200 g/mol
Melting Pt: 61…62 °C
Storage Temperature: Ambient
MDL Number: MFCD11617316
CAS Number: 931105-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR