Methyl-6-[4-(trifluoromethyl)piperidino]nicotinate
Supplier: Apollo Scientific
PC200249-1G
PC200249-500MG
APOSPC200249-1GEA
248
EUR
APOSPC200249-1G
APOSPC200249-500MG
Methyl-6-[4-(trifluoromethyl)piperidino]nicotinate
Methyl-6-[4-(trifluoromethyl)piperidino]nicotinate
Formula:
C₁₃H₁₅F₃N₂O₂ MW: 288,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10758065 CAS Number: 1033463-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...