3-Bromo-5-chloro-6-(trifluoromethyl)-1H-indole

Supplier: Apollo Scientific

PC200257-1G PC200257-500MG
APOSPC200257-1GEA 519 EUR
APOSPC200257-1G APOSPC200257-500MG
3-Bromo-5-chloro-6-(trifluoromethyl)-1H-indole
3-Bromo-5-chloro-6-(trifluoromethyl)-1H-indole
Formula: C₉H₄BrClF₃N
Melting Pt: 89…91 °C
Storage Temperature: Ambient
MDL Number: MFCD12827786
CAS Number: 1186404-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR