5-Bromo-2-(trifluoromethyl)pyrimidine 96%
Supplier: Apollo Scientific
|
Warning
|
PC200259-1G
PC200259-10G
PC200259-5G
APOSPC200259-1GEA
74.2
EUR
APOSPC200259-1G
APOSPC200259-10G
APOSPC200259-5G
5-Bromo-2-(trifluoromethyl)pyrimidine 96%
5-Bromo-2-(trifluoromethyl)pyrimidine
Formula:
C₅H₂BrF₃N₂ MW: 226,98 g/mol Melting Pt: 41…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD09754046 CAS Number: 799557-86-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...