tert-Butyl-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}tetrahydro-1(2H)-pyridinecarboxylate
Supplier: Apollo Scientific
PC200270-1G
PC200270-500MG
APOSPC200270-1GEA
364
EUR
APOSPC200270-1G
APOSPC200270-500MG
tert-Butyl-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}tetrahydro-1(2H)-pyridinecarboxylate
tert-Butyl-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}tetrahydro-1(2H)-pyridinecarboxylate
Formula:
C₁₆H₂₁ClF₃N₃O₂ MW: 379,81 g/mol Melting Pt: 95…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD11553061 CAS Number: 1092352-86-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...