4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrazol-3-amine 95%
Supplier: Apollo Scientific
|
Warning
|
PC200307-500MG
PC200307-1G
APOSPC200307-500MGEA
510
EUR
APOSPC200307-500MG
APOSPC200307-1G
4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrazol-3-amine 95%
4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrazol-3-amine
Formula:
C₉H₆ClF₃N₄ Melting Pt: 167…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD16622823 CAS Number: 1242267-93-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...