Ethyl-3-amino-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Supplier: Apollo Scientific
|
Warning
|
PC200327-500MG
PC200327-1G
APOSPC200327-500MGEA
528
EUR
APOSPC200327-500MG
APOSPC200327-1G
Ethyl-3-amino-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Ethyl 3-amino-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Formula:
C₁₁H₁₀F₃N₃O₂ Melting Pt: 219…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD12498719 CAS Number: 1171919-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...