(Morpholinium-4-ylmethyl)trifluoroborate internal salt 95%

Supplier: Apollo Scientific
Warning

PC200335-1G PC200335-500MG
APOSPC200335-1GEA 248 EUR
APOSPC200335-1G APOSPC200335-500MG
(Morpholinium-4-ylmethyl)trifluoroborate internal salt 95%
(Morpholinium-4-ylmethyl)trifluoroborate internal salt
Formula: C₅H₁₁BF₃NO
MW: 168,95 g/mol
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD20134168
CAS Number: 1268340-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR