3,4-Dibromo-2-fluoropyridine
Supplier: Apollo Scientific
|
Warning
|
PC200341-1G
PC200341-500MG
APOSPC200341-1GEA
364
EUR
APOSPC200341-1G
APOSPC200341-500MG
3,4-Dibromo-2-fluoropyridine
3,4-Dibromo-2-fluoropyridine
Formula:
C₅H₂Br₂FN Melting Pt: 74…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD09037460 CAS Number: 137718-84-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...