6-Chloro-2-fluoronicotinic acid 97%

Supplier: Apollo Scientific

Synonyms: 6-Chloro-2-fluoro-3-pyridinecarboxylic acid

PC201063-100MG PC201063-1G PC201063-250MG
APOSPC201063-100MGEA 96 EUR
APOSPC201063-100MG APOSPC201063-1G APOSPC201063-250MG
6-Chloro-2-fluoronicotinic acid 97%
6-Chloro-2-fluoronicotinic acid
Formula: C₆H₃ClFNO₂
MW: 175,543 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16610397
CAS Number: 1211578-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR