3-Chloro-5-fluoropyridin-2-amine 97%
Supplier: Apollo Scientific
PC201069-1G
PC201069-250MG
PC201069-5G
APOSPC201069-1GEA
65.7
EUR
APOSPC201069-1G
APOSPC201069-250MG
APOSPC201069-5G
3-Chloro-5-fluoropyridin-2-amine 97%
3-Chloro-5-fluoropyridin-2-amine
Formula:
C5H4ClFN2 Storage Temperature: Ambient |
MDL Number:
MFCD09260907 CAS Number: 1214330-79-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...