5-Bromo-2-fluoronicotinonitrile 95%

Supplier: Apollo Scientific

PC201096-100MG PC201096-1G PC201096-250MG
APOSPC201096-100MGEA 87 EUR
APOSPC201096-100MG APOSPC201096-1G APOSPC201096-250MG
5-Bromo-2-fluoronicotinonitrile 95%
5-Bromo-2-fluoronicotinonitrile
Formula: C₆H₂BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD18258637
CAS Number: 1256821-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR