4-Amino-2-(trifluoromethyl)pyridine 98%
Supplier: Apollo Scientific
|
Danger
|
PC201143-5G
PC201143-1G
APOSPC201143-5GEA
71.3
EUR
APOSPC201143-5G
APOSPC201143-1G
4-Amino-2-(trifluoromethyl)pyridine 98%
4-Amino-2-(trifluoromethyl)pyridine
Formula:
C₆H₅F₃N₂ MW: 162,1100 g/mol Melting Pt: 58…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD09056877 CAS Number: 147149-98-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...