((Difluoromethyl)sulphonyl)benzene 98%

Supplier: Apollo Scientific

Synonyms: Difluoromethyl phenyl sulphone

PC201148-100G PC201148-25G
APOSPC201148-100GEA 600 EUR
APOSPC201148-100G APOSPC201148-25G
((Difluoromethyl)sulphonyl)benzene 98%
((Difluoromethyl)sulphonyl)benzene
Formula: C₇H₆F₂O₂S
MW: 192,18 g/mol
MDL Number: MFCD01050170
CAS Number: 1535-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR