2,6-Difluoro-4-formylbenzonitrile 98%
Supplier: Apollo Scientific
PC201253-1G
PC201253-25G
PC201253-5G
APOSPC201253-1GEA
82.9
EUR
APOSPC201253-1G
APOSPC201253-25G
APOSPC201253-5G
2,6-Difluoro-4-formylbenzonitrile 98%
2,6-Difluoro-4-formylbenzonitrile
Formula:
C₈H₃F₂NO MW: 167,1100 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09263470 CAS Number: 433939-88-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...