6-Fluoro-1-tetralone 97%
Supplier: Apollo Scientific
PC201285-1G
PC201285-5G
APOSPC201285-1GEA
98.6
EUR
APOSPC201285-1G
APOSPC201285-5G
6-Fluoro-1-tetralone 97%
6-Fluoro-1-tetralone
Formula:
C₁₀H₉FO MW: 164,1763 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09031370 CAS Number: 703-67-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...