Ethyl-4-amino-3-fluorobenzoate 97%

Supplier: Apollo Scientific

Synonyms: Ethyl 4-amino-3-fluorobenzoate

PC201297-10G PC201297-5G
APOSPC201297-10GEA 133 EUR
APOSPC201297-10G APOSPC201297-5G
Ethyl-4-amino-3-fluorobenzoate 97%
Ethyl 4-amino-3-fluorobenzoate
Formula: C₉H₁₀FNO₂
MW: 183,1796 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782043
CAS Number: 73792-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR