5-Bromo-4-(trifluoromethyl)pyrimidine 97%

Supplier: Apollo Scientific

PC201307-250MG PC201307-1G
APOSPC201307-250MGEA 82.9 EUR
APOSPC201307-250MG APOSPC201307-1G
5-Bromo-4-(trifluoromethyl)pyrimidine 97%
5-Bromo-4-(trifluoromethyl)pyrimidine
Formula: C₅H₂BrF₃N₂
MW: 226,9820 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11046325
CAS Number: 785777-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR