5-Bromo-6-trifluoromethylpyridin-2-ylamine 95+%

Supplier: Apollo Scientific

PC201346-10G PC201346-1G PC201346-5G
APOSPC201346-10GEA 344 EUR
APOSPC201346-10G APOSPC201346-1G APOSPC201346-5G
5-Bromo-6-trifluoromethylpyridin-2-ylamine 95+%
5-Bromo-6-trifluoromethylpyridin-2-ylamine
Formula: C₆H₄BrF₃N₂
MW: 241,0086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848010
CAS Number: 882500-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR