2-Chloro-3-(trifluoromethyl)pyridine 98%
Supplier: Apollo Scientific
|
Danger
|
PC2075-100G
PC2075-50G
APOSPC2075-100GEA
72.7
EUR
APOSPC2075-100G
APOSPC2075-50G
2-Chloro-3-(trifluoromethyl)pyridine 98%
2-Chloro-3-(trifluoromethyl)pyridine
Formula:
C₆H₃ClF₃N MW: 181,54 g/mol Boiling Pt: 166…168 °C Melting Pt: 36…40 °C Flash Pt: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042223 CAS Number: 65753-47-1 EINECS: 424-520-6 UN: 2923 ADR: 8+6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...