2-Chloro-5-fluoronicotinonitrile 95%

Supplier: Apollo Scientific

PC210031-1G PC210031-500MG PC210031-5G PC210031-25G
APOSPC210031-1GEA 63.8 EUR
APOSPC210031-1G APOSPC210031-500MG APOSPC210031-5G APOSPC210031-25G
2-Chloro-5-fluoronicotinonitrile 95%
2-Chloro-5-fluoronicotinonitrile
Formula: C₆H₂ClFN₂
Storage Temperature: Ambient
MDL Number: MFCD15526908
CAS Number: 791644-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR