1-(2-Cyanoethyl)-5-(trifluoromethyl)pyrid-2-(1H)-one
Supplier: Apollo Scientific
|
Warning
|
PC2108-1G
PC2108-10G
APOSPC2108-1GEA
29
EUR
APOSPC2108-1G
APOSPC2108-10G
1-(2-Cyanoethyl)-5-(trifluoromethyl)pyrid-2-(1H)-one
1-(2-Cyanoethyl)-5-(trifluoromethyl)pyrid-2-(1H)-one
Formula:
C₉H₇F₃N₂O MW: 216,16 g/mol Melting Pt: 89…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00204218 CAS Number: 175277-71-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...