4-Benzyloxybenzotrifluoride 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-(Benzyloxy)benzotrifluoride
PC2161-1G
PC2161-250MG
APOSPC2161-1GEA
120
EUR
APOSPC2161-1G
APOSPC2161-250MG
4-Benzyloxybenzotrifluoride 98%
4-Benzyloxybenzotrifluoride
Formula:
C₁₄H₁₁F₃O MW: 252.23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00275905 CAS Number: 70097-65-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...