1,6-Dibromoperfluorohexane 96%

Supplier: Apollo Scientific
Warning

Synonyms: Perfluoro(1,6-dibromohexane)

PC2253H-25G PC2253H-5G PC2253H-1G
APOSPC2253H-25GEA 287 EUR
APOSPC2253H-25G APOSPC2253H-5G APOSPC2253H-1G
1,6-Dibromoperfluorohexane 96%
1,6-Dibromoperfluorohexane
Formula: C₆Br₂F₁₂
MW: 459,87 g/mol
Boiling Pt: 141…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00153110
CAS Number: 918-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR