4-Bromo-3-fluorobenzenesulphonyl chloride 98%
Supplier: Apollo Scientific
|
Danger
|
PC2306-100G
PC2306-25G
PC2306-5G
APOSPC2306-100GEA
259
EUR
APOSPC2306-100G
APOSPC2306-25G
APOSPC2306-5G
4-Bromo-3-fluorobenzenesulphonyl chloride 98%
4-Bromo-3-fluorobenzenesulphonyl chloride
Formula:
C₆H₃BrClFO₂S Boiling Pt: 217…218 °C Density: 1,85 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094150 CAS Number: 351003-51-5 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...