3,4-Dichloro-α,α,α-trifluorotoluene 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1,2-Dichloro-4-(trifluoromethyl)benzene, 3,4-Dichlorobenzotrifluoride
PC2340-100G
PC2340-500G
APOSPC2340-100GEA
14.7
EUR
APOSPC2340-100G
APOSPC2340-500G
3,4-Dichloro-α,α,α-trifluorotoluene 98%
3,4-Dichloro-α,α,α-trifluorotoluene
Formula:
C₇H₃Cl₂F₃ MW: 215 g/mol Boiling Pt: 173…174 °C Melting Pt: –12 °C Density: 1,479 g/cm³ (25 °C) Flash Pt: 65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000555 CAS Number: 328-84-7 EINECS: 206-337-1 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...