2-Fluoronaphthalene
Supplier: Apollo Scientific
|
Danger
|
PC2378-1G
PC2378-5G
APOSPC2378-1GEA
198
EUR
APOSPC2378-1G
APOSPC2378-5G
2-Fluoronaphthalene
2-Fluoronaphthalene
Formula:
C₁₀H₇F MW: 146,16 g/mol Boiling Pt: 90 °C/16mmHg Melting Pt: 61 °C Flash Pt: 65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082618 CAS Number: 323-09-1 EINECS: 206-299-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...