1,3-Dichloro-5-fluorobenzene 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3,5-Dichlorofluorobenzene
PC2401-100G
PC2401-25G
APOSPC2401-100GEA
376
EUR
APOSPC2401-100G
APOSPC2401-25G
1,3-Dichloro-5-fluorobenzene 99%
1,3-Dichloro-5-fluorobenzene
Formula:
C₆H₃Cl₂F MW: 164,9900 g/mol Boiling Pt: 162 °C Density: 1,37 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00060652 CAS Number: 1435-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...