3-Bromo-4'-fluorobiphenyl 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Bromo-4'-Fluoro-1,1'-biphenyl
PC2411-1G
PC2411-5G
APOSPC2411-1GEA
260
EUR
APOSPC2411-1G
APOSPC2411-5G
3-Bromo-4'-fluorobiphenyl 97%
3-Bromo-4'-fluorobiphenyl
Formula:
C₁₂H₈BrF MW: 251.0943 g/mol Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD04038228 CAS Number: 10540-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...