Methyl-2-(trifluoromethyl)benzoylacetate 95+%

Supplier: Apollo Scientific

PC2418-1G PC2418-5G PC2418-25G
APOSPC2418-1GEA 143 EUR
APOSPC2418-1G APOSPC2418-5G APOSPC2418-25G
Methyl-2-(trifluoromethyl)benzoylacetate 95+%
Methyl-2-(trifluoromethyl)benzoylacetate
Formula: C₁₁H₉F₃O₃
MW: 246,19 g/mol
Boiling Pt: 98…105 °C/0,41mmHg
Storage Temperature: Ambient
MDL Number: MFCD00216523
CAS Number: 212755-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR