3-Bromo-4-fluorophenylacetic acid 98%
Supplier: Apollo Scientific
|
Warning
|
PC2441-100G
PC2441-25G
PC2441-5G
APOSPC2441-100GEA
154
EUR
APOSPC2441-100G
APOSPC2441-25G
APOSPC2441-5G
3-Bromo-4-fluorophenylacetic acid 98%
3-Bromo-4-fluorophenylacetic acid
Formula:
C₈H₆BrFO₂ MW: 233,04 g/mol Melting Pt: 65…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD04038373 CAS Number: 194019-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...