2-Bromo-3-methoxy-1,1,1-trifluoropropane 97%
Supplier: Apollo Scientific
|
Warning
|
PC2485-25G
PC2485-5G
APOSPC2485-25GEA
876
EUR
APOSPC2485-25G
APOSPC2485-5G
2-Bromo-3-methoxy-1,1,1-trifluoropropane 97%
2-Bromo-3-methoxy-1,1,1-trifluoropropane
Formula:
C₄H₆BrF₃O Storage Temperature: Ambient |
MDL Number:
MFCD04038268 CAS Number: 883498-92-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...