2-Chloro-5-fluoropyridine 99%
Supplier: Apollo Scientific
|
Warning
|
PC2546-100G
PC2546-25G
PC2546-5G
APOSPC2546-100GEA
168
EUR
APOSPC2546-100G
APOSPC2546-25G
APOSPC2546-5G
2-Chloro-5-fluoropyridine 99%
2-Chloro-5-fluoropyridine
Formula:
C₅H₃ClFN Boiling Pt: 74 °C/38mmHg Melting Pt: 19 °C Density: 1,297 g/cm³ (25 °C) Flash Pt: 42 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095332 CAS Number: 31301-51-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...