2',6'-Difluoroacetophenone 96%

Supplier: Apollo Scientific
Warning

PC2577-5G PC2577-100G PC2577-25G
APOSPC2577-5GEA 21.9 EUR
APOSPC2577-5G APOSPC2577-100G APOSPC2577-25G
2',6'-Difluoroacetophenone 96%
2',6'-Difluoroacetophenone
Formula: C₈H₆F₂O
Boiling Pt: 76…79 °C/15mmHg
Density: 1,195 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00000328
CAS Number: 13670-99-0
EINECS: 237-151-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR