3',4'-Difluoroacetophenone 97%

Supplier: Apollo Scientific
Warning

Synonyms: 1-(3,4-Difluorophenyl)ethan-1-one, 1-(3,4-Difluorophenyl)ethanone

PC2578-100G PC2578-25G PC2578-500G
APOSPC2578-100GEA 49.5 EUR
APOSPC2578-100G APOSPC2578-25G APOSPC2578-500G
3',4'-Difluoroacetophenone 97%
3',4'-Difluoroacetophenone
Formula: C₈H₆F₂O
MW: 156,13 g/mol
Boiling Pt: 94…95 °C/13mmHg
Melting Pt: 19…20 °C
Density: 1,246 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00009891
CAS Number: 369-33-5
EINECS: 206-717-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR