1-Iodo-4-(1,1,2,2-tetrafluoroethoxy)benzene
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Iodo-alpha,alpha,beta,beta-tetrafluorophenetole
PC2601-1G
PC2601-5G
PC2601-10G
APOSPC2601-1GEA
79.9
EUR
APOSPC2601-1G
APOSPC2601-5G
APOSPC2601-10G
1-Iodo-4-(1,1,2,2-tetrafluoroethoxy)benzene
1-Iodo-4-(1,1,2,2-tetrafluoroethoxy)benzene
Formula:
C₈H₅F₄IO Storage Temperature: Ambient |
CAS Number:
863323-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...