3,5-Difluoroanisole 98%

Supplier: Apollo Scientific
Warning

Synonyms: 1,3-Difluoro-5-methoxybenzene

PC2614J-100G PC2614J-25G PC2614J-5G
APOSPC2614J-100GEA 144 EUR
APOSPC2614J-100G APOSPC2614J-25G APOSPC2614J-5G
3,5-Difluoroanisole 98%
3,5-Difluoroanisole
Formula: C₇H₆F₂O
MW: 144,12 g/mol
Boiling Pt: 145…148 °C
Density: 1,234 g/cm³ (25 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00042560
CAS Number: 93343-10-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR