3,5-Difluorobenzamide 97%
Supplier: Apollo Scientific
|
Warning
|
PC2619D-100G
PC2619D-25G
PC2619D-5G
APOSPC2619D-100GEA
173
EUR
APOSPC2619D-100G
APOSPC2619D-25G
APOSPC2619D-5G
3,5-Difluorobenzamide 97%
3,5-Difluorobenzamide
Formula:
C₇H₅F₂NO MW: 157.12 g/mol Melting Pt: 155…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061138 CAS Number: 132980-99-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...