Tris(2,2,2-trifluoroethyl) Phosphate

Supplier: Apollo Scientific
Warning

Synonyms: Tris(2,2,2-trifluoroethyl)phosphate, Phosphoric acid tris(2,2,2-trifluoroethyl) ester

PC2653-25G PC2653-5G PC2653-100G
APOSPC2653-25GEA 466 EUR
APOSPC2653-25G APOSPC2653-5G APOSPC2653-100G
Tris(2,2,2-trifluoroethyl) Phosphate
Tris(2,2,2-trifluoroethyl) Phosphate
Formula: C₆H₆F₉O₄P
MW: 344,07 g/mol
Boiling Pt: 83…86 °C/11mmHg
Melting Pt: –20 °C
Density: 1,5941 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00233117
CAS Number: 358-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR