3,4-Difluorobenzophenone 98%

Supplier: Apollo Scientific
Warning

PC2694-10G PC2694-50G PC2694-25G PC2694-100G
APOSPC2694-10GEA 21.9 EUR
APOSPC2694-10G APOSPC2694-50G APOSPC2694-25G APOSPC2694-100G
3,4-Difluorobenzophenone 98%
3,4-Difluorobenzophenone
Formula: C₁₃H₈F₂O
Melting Pt: 53…55 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00009892
CAS Number: 85118-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR