2,6-Difluorobenzoyl chloride 98%

Supplier: Apollo Scientific
Danger

PC2725-100G PC2725-250G
APOSPC2725-100GEA 118 EUR
APOSPC2725-100G APOSPC2725-250G
2,6-Difluorobenzoyl chloride 98%
2,6-Difluorobenzoyl chloride
Formula: C₇H₃ClF₂O
MW: 176,55 g/mol
Boiling Pt: 72…75 °C/13mmHg
Density: 1,404 g/cm³ (25 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00000659
CAS Number: 18063-02-0
EINECS: 241-971-2
UN: 2920
ADR: 8+3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR