N-[3-(Trifluoromethyl)benzyl]urea 95%

Supplier: Apollo Scientific

PC28174-1G PC28174-5G
APOSPC28174-1GEA 180 EUR
APOSPC28174-1G APOSPC28174-5G
N-[3-(Trifluoromethyl)benzyl]urea 95%
N-[3-(Trifluoromethyl)benzyl]urea
Formula: C₉H₉F₃N₂O
MW: 218,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01314643
CAS Number: 296277-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR