3-(4-Chlorophenyl)-2-fluoro-3-oxopropanenitrile 95%

Supplier: Apollo Scientific

PC28203-1G PC28203-5G
APOSPC28203-1GEA 299 EUR
APOSPC28203-1G APOSPC28203-5G
3-(4-Chlorophenyl)-2-fluoro-3-oxopropanenitrile 95%
3-(4-Chlorophenyl)-2-fluoro-3-oxopropanenitrile
Formula: C₉H₅ClFNO
Storage Temperature: Ambient
MDL Number: MFCD18426510
CAS Number: 1263063-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR